Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

Cover glasses

Deckgläser, 0,17 ±0,02 mm, 18 mm
  HECH92101501080
Cover glasses
  HECH92101501080
Cover glasses
Cover glasses

Deckgläser, 0,17 ±0,02 mm, 18 mm

 
 
   
1 * 1.000 ST
 

High spectral transmission, excellent flatness, optimum resistance to chemicals.

  • Free from colour distortion borosilicate glass D 263™ M
  • Excellent quality: Low levels of inclusions, striae, bubbles and streaks

 : ISO 8255

 : In tight-closing plastic boxes for protection against damp and climatic influences.

 
  • Ø:
    18 mm
  • Bezeichnung:
    Deckgläser
  • Seq:
    03
  • Stärke:
    0,17 ±0,02 mm