AuftragsverfolgungKontakt
Blood collection needles, BD Vacutainer® Eclipse™ | Avantor
Blood collection needles, BD Vacutainer® Eclipse™
21, 32 mm, Grün
Katalognr. 613-3110
Blood collection needles, BD Vacutainer® Eclipse™
Katalognr. 613-3110
Anbieter:  
Blood collection needles, BD Vacutainer® Eclipse™
Blood collection needles, BD Vacutainer® Eclipse™
21, 32 mm, Grün
Artikel gestrichen
Das Produkt wurde eingestellt. Wir haben jedoch ähnliche Alternativprodukte gefunden.
Alternativen
KANÜLE VACUTAINER ECLIPSE 21G 1,25INCH 1 * 48 ST Katalognr.BDAM368609
Das Produkt BDAM368609 ist ein Ersatz für 613-3110, variiert jedoch in Marke, Verpackung oder Menge.
Product Details & Documents
The BD Vacutainer® Eclipse™ blood collection needle is a safety-engineered, multi-sample blood collection needle that offers a simple, effective way to collect blood while reducing the possibility of needlestick injuries. It features a safety shield that allows for one-handed activation to cover the needle immediately upon withdrawal from the vein and confirms proper activation with an audible click.
  • Built-in safety shield is aligned with the bevel of the needle
  • The safety shield permanently locks into place, significantly reducing the risk of needlestick injuries
  • The pre-attached holder (only supplied with some models) minimises the risk of accidental needlestick injuries from the holder end of the device
Technische Daten
  • Farbcode:
    Grün
  • Gauge:
    21
  • Länge:
    32 mm
  • Seq:
    02