Storage bags
Storage bags, transparent, with ground weld., 0,05 mm, 100×200 mm
Catalog # SEMA2442
Supplier: SEMADENI
Jump to:
Product Details & Documents
Storage bags in transparent LDPE, with bottom weld.
It has bottom weld. They are also suitable for liquids but without side weld.
Packaging: Minimum quantity:100 pieces
Specifications
- Bezeichnung:Storage bags, transparent, with ground weld.
- Abmessungen:100×200 mm
- Seq:03
- Stärke:0,05 mm