Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Racks for freezer

Racks for freezer

12 (1×12), 32 mm, 140×140×443 mm
Katalognr. RATI5400033
Anbieter:   RATIOLAB
Racks for freezer
Katalognr. RATI5400033
Anbieter:  RATIOLAB
Racks for freezer

12 (1×12), 32 mm, 140×140×443 mm

Preis
SFr. 198.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

For the storing of cryoboxes in freezer cabinets and chests.

  • Robust construction
  • Handles for convenient transport
  • Made of stainless steel with surfaces for perfect cleaning

Delivery information: Cryo racks are supplied with shelf sizes for any standard cryoboxes.

Technische Daten
  • Fächer:
    12 (1×12)
  • Außen B×T×H:
    140×140×443 mm
  • Seq:
    11
  • Kryoboxhöhe:
    32 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.