Conductive pipette tips
Conductive, slim style, 10 - 240 µl, -, -, Schwarz, Bulk, 900/bag
Katalognr. GREI764280_7200
Anbieter: Greiner Bio-One
Direkt zu
Product Details & Documents
Conductive tips are used to prevent carryover in automated pipetting robots. The addition of graphite to the raw material PP makes the pipette tips electrically conductive. By measuring electric currents, the filling level can be determined in sample and reagent containers, so that the depth of immersion of the tip can be adjusted to the filling level.
- Filter tip: Free of detectable DNase, RNase, human DNA; non pyrogenic, non cytotoxic
Technische Daten
- Verpackt:Bulk, 900/bag
- Steril:-
- Volumen:10 - 240 µl
- Spitzentyp:Conductive, slim style
- Spitzenfarbe:Schwarz
- Filter:-
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.