Water purification systems, Select Analyst
Select Analyst 40 GP with boost pump, without internal tank (external tank must be ordered separately)
171-0276
: Purite
The Select Analyst is a compact unit ideal for Laboratory use, delivering a steady supply of > 1MΩ.cm pure water for Laboratory tasks using carbon pre-treatment, reverse osmosis and deionisation.
- RO removes > 98% minerals and > 99% bacteria
- RO boost pump fitted as standard
- LCD colour touch screen panel, visual and audible alarms
- Available with or without integral 20 litre storage
- Semi-automatic clean cycle
- USB port to download event data and upload software updates
- WiFi connection
Typical applications include glassware rinsing, buffers and stains, reagent make-up, and media preparation.
Installation kit and all consumables included for first year's operation.
: Supplied with alphanumeric display panel, pretreatment cartridge, reverse osmosis module, deionisation module and single tank bib-tap. External tanks required for GP models are also available, please contact your local Avantor office for details.
- Bezeichnung:Select Analyst 40 GP with boost pump, without internal tank (external tank must be ordered separately)
- Seq:01
- Anorganische Stoffe @ 25°C:>1 MΩ/cm
- Produktionsfließrate:3,6 - 6 L/h
- Cat. No.:171-0276
- TOC-Gehalt:<50 ppb
- Modell:Analyst 40
- Bakterien:>99% rejection*
- B×T×H:440×560×750 mm
- Speisewasser:Potable (tap)
- Gewicht:28 - 43 kg
Accessories
- Bezeichnung
- 171-0614External tank wall mounting kit, for Select Analyst and HP systems1 * 1 ST - SFr. 1’790.00
- Bezeichnung
- Bezeichnung
- 171-0256Jährliches Verbrauchsmaterialpaket für Select Analyst 320/Purewater 300 mit 2 x PT16- und 2 x PP16-Kartuschen1 * 1 ST - SFr. 5’260.00
- Bezeichnung
- 171-0265Filterelement, 0,2 µm für die aktuelle Select-Baureihe (mit grauer Tür) und vorheriger L2G Select-Baureihe (mit blauer Tür), werden in HP-, Fusion-, Neptun-Wasseraufbereitungssystemen verwendet1 * 1 ST - SFr. 981.00
- Bezeichnung



