Pipette tips
QuickRack, 1 - 200 µl, Gelb
Katalognr. SORE30460
Anbieter: Sorenson Bioscience
Direkt zu
Product Details & Documents
These universal fit pipette tips are manufactured from medical grade polypropylene in a tightly controlled production environment.
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
The accurate and precise tips are designed to fit most research-grade pipettors. Available in low binding, pipette tips reduce sample retention.
Technische Daten
- Volumen:1 - 200 µl
- Seq:03
- Spitzentyp:QuickRack
- Spitzenfarbe:Gelb