AuftragsverfolgungKontakt
One-Step SYBR® Green qRT-PCR, qScript™
One-Step SYBR® Green qRT-PCR, qScript™
qScript™ One-Step SYBR® Green qRT-PCR Kit, with low ROX, 200×50 µl reactions, ABI 7500/7500 Fast/QuantStudio™, Agilent AriaMx, Douglas Scientific IntelliQube®, Stratagene Mx, 200 Assays
Katalognr. 733-2082
Anbieter:   Quantabio
One-Step SYBR® Green qRT-PCR, qScript™
Katalognr. 733-2082
Anbieter:  Quantabio
CAS Nummer:  
One-Step SYBR® Green qRT-PCR, qScript™
qScript™ One-Step SYBR® Green qRT-PCR Kit, with low ROX, 200×50 µl reactions, ABI 7500/7500 Fast/QuantStudio™, Agilent AriaMx, Douglas Scientific IntelliQube®, Stratagene Mx, 200 Assays
CAS Nummer:  
Preis
804.00 SFr.
Einloggen für Verfügbarkeit
Product Details & Documents
qScript™ One-Step SYBR® Green qRT-PCR Kits reliably deliver maximum qRT-PCR efficiency, sensitivity and specificity in with a simplified protocol. These kits offer reliable and unbiased amplification of all RNA targets regardless of abundance. The proprietary supermix maximises activities of both reverse transcriptase and AccuStart™ Taq DNA polymerase without compromising specificity of amplification. qScript™ One-Step SYBR® Green qRT-PCR kits are available and optimised for all real-time PCR instrument platforms including those requiring normalisation with ROX reference dye.
Delivery information: Kits are available with or without ROX or with fluorescein for Bio-Rad iCycler iQ® systems. All kits contain qScript One-Step Reverse Transcriptase (optimised 50X formulation of recombinant MMLV reverse transcriptase for one-step RT-PCR); One-Step SYBR Green Master Mix (2X) reaction buffer containing dNTPs, MgCl₂, AccuStart™ Taq DNA polymerase, stabilisers, and SYBR® Green I dye; and nuclease-free water.
Technische Daten
Technische Daten
  • Cycler-Kompatibilität:
    ABI 7500/7500 Fast/QuantStudio™, Agilent AriaMx, Douglas Scientific IntelliQube®, Stratagene Mx
  • VE:
    200 Assays
  • Bezeichnung:
    qScript™ One-Step SYBR® Green qRT-PCR Kit, with low ROX, 200×50 µl reactions
  • Anwendung:
    RT-qPCR dye
  • Cat. No.:
    733-2082
  • One step:
    Ja