Buffer solutions, according to DIN 19266, Fluka™
6,865, —, Phosphat, 25 °C, Plastic bottle, 6×1 L, 6 L
Katalognr. 33594-6X1L
Anbieter: Honeywell Chemicals
Buffer solutions, according to DIN 19266, Fluka™
Katalognr. 33594-6X1L
Anbieter: Honeywell Chemicals
CAS Nummer:
Direkt zu
Product Details & Documents
Buffers with fungicide (except pH 10,012)
Zertifizierung: According to DIN 19266.
Dokumente
Technische Daten
Technische Daten
- Packung:Plastic bottle
- pH-Puffertyp:Phosphat
- VE:6 L
- Referenztemperatur:25 °C
- pH Genauigkeit:—
- Pk Info:6×1 L
- pH-Wert:6,865



