Ethyltricosanoat
Glasflasche für Feststoffe, 10 mg
Katalognr. CAYM10006874-10
Anbieter: Cayman Chemical
Direkt zu
Product Details & Documents
Synonyme: Tricosanoic acid ethyl ester
- CAS Nummer:18281-07-7
- Formel:C₂₅H₅₀O₂
- Molekulargewicht:382.67 g/mol
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:10 mg
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



