Single channel pipettes, variable volume
100 - 1000 µl
Katalognr. 613-1362
Anbieter: EDVOTEK
Direkt zu
Product Details & Documents
Sturdily designed handheld pipettes with volumes ranging from 0,1 to 5000 µl. They are easy to use, accurate and use standard micropipette tips.
- Volume easily selected by twisting the top
- Lightweight design and tip ejector makes operation fast and easy
- Tool and instructions included for self-calibration
Technische Daten
- Volumen:100 - 1000 µl