Gewebehomogenisatoren Dounce, KIMBLE®
2 ml
Katalognr. 432-0254
Anbieter: DWK Life Sciences
Direkt zu
Product Details & Documents
Designed primarily for cellular work where the nucleus remains intact after homogenisation.
- All-glass construction
- Two pestles are supplied with each complete unit
- Large clearance pestle is used for the initial sample reduction
- Small clearance pestle is used to form the final homogenate
- Replacement components are available and completely interchangeable
Technische Daten
- Kapazität:2 ml
- Seq:14