Drum taps, brass
Self-closing with flame guard and PTFE seal, Viscous liquids >30 SAE, 2”
Katalognr. MSGA2800
Anbieter: asecos
Direkt zu
Product Details & Documents
Brass taps for drums with screw thread.
- Self closing
- Flame guard
- PTFE seal
Ordering information: MSGA2799 includes a trailer fitting for decanting vessels.
Technische Daten
- Gewinde:2”
- Bezeichnung:Self-closing with flame guard and PTFE seal
- Seq:03
- Für:Viscous liquids >30 SAE