Absorption granules - Universal coarse grain
Universal Coarse Grain absorption granules, 1 - 3 mm, 16 litres/sack, 20 kg/sack
Katalognr. MSGA9956
Anbieter: asecos
Direkt zu
Product Details & Documents
Absorption granules for oil, alkalis, grease, motor fuels, varnishes.
- For outdoor and indoor use
- Officially tested and approved for use on traffic areas
- 20 kg sack binds up to 16 litres of heating oil
- Suitable for solid surfaces
- Even with total saturation of the granules, anti-slipping is guaranteed
Technische Daten
- Bezeichnung:Universal Coarse Grain absorption granules, 1 - 3 mm
- Absorptionsvermögen:16 litres/sack
- Verpackt:20 kg/sack