Calibrated Baumé hydrometers
–2 - 15, 0,5, 300 mm
Katalognr. ALLA0151FC002/20QP
Anbieter: Alla France
Direkt zu
Product Details & Documents
Baumé hydrometers calibrated at 20 °C.
Technische Daten
- Länge:300 mm
- Seq:02
- Teilung:0,5
- Messbereich:–2 - 15
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



