Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Calibrated Baumé hydrometers

Calibrated Baumé hydrometers

–2 - 10, 0,1, 315 mm
Catalog # ALLA0101TC002/20QP
Supplier:   Alla France
Calibrated Baumé hydrometers
Catalog # ALLA0101TC002/20QP
Supplier:  Alla France
Calibrated Baumé hydrometers
Calibrated Baumé hydrometers

–2 - 10, 0,1, 315 mm

Price
SFr. 130.00
Log in for availability
1 * 1 ST
Product Details & Documents

Baumé hydrometers calibrated at 20 °C.

Specifications
  • Länge:
    315 mm
  • Seq:
    05
  • Teilung:
    0,1
  • Messbereich:
    –2 - 10