Automation tips for Tecan®, Axygen®
Product Details & Documents
PP, mit oder ohne Filter, steril oder unsteril
- Spitzen mit Füllstandssensor
- Optimierte feine Spitze für genaues Dispensieren
- Als frei von RNase, DNase und Endotoxinen zertifiziert
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
EVO-style tips are compatible with Tecan Freedom EVO®, Caliper Zephyr® SPE, and BioTek® Precision™ robotic pipetting systems.
Genesis-style tips are compatible with Tecan Genesis Freedom, Freedom EVO, and MiniPrep robotic workstations, as well as Cavro®, PerkinElmer/Packard Multiprobe II HT, and Multiprobe II HT EX workstations.
Packaging: Packaged in plastic blister packs.
Caution: * Economy-Verpackung in einem Karton
Documents
Specifications
- Verpackt:50 Racks mit je 96 Stück
- Steril:+
- Volumen:50 µl
- Seq:27
- Spitzentyp:EVF-50-R-S
- Spitzenfarbe:Transparent
- Zur Nutzung mit:MCA96 / HV96
- Filter:+