pH-Kombinationselektroden
Sensolyt® EC*, Sensolyt® EC, ohne Armierung, zur Installation in Durchflussgefäße, 2 - 12, Normal kontaminiertes Wasser
Katalognr. WTWO109102
Anbieter: WTW ONLINE
Direkt zu
Product Details & Documents
The SensoLyt® measuring chains with arming are easy to install and boast flexible measuring and application areas. Eletrodes without arming can be connected directly to a pH monitor of the EcoLine® or QuadroLine® series. Depending on the application, they can be installed into a flow vessel.
- Ideal for measurements in water and drinking water
- Also for high pressure and temperature conditions
- Easy to maintain by easy measuring chain replacement
Technische Daten
- Für:Normal kontaminiertes Wasser
- Typ:Sensolyt® EC, ohne Armierung, zur Installation in Durchflussgefäße
- Bezeichnung:Sensolyt® EC*
- Seq:32
- pH:2 - 12