Multidishes, Nunc™ UpCell™ surface
6-Well-Multischale, UpCell™, belüftet, 9,6 cm², 3,0 ml
Catalog # 734-2252
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
The UpCell™ surface is a temperature-responsive cell culture surface that enables harvesting of cells with high viability and intact surface proteins for culture passaging, single-cell analyses and cell transplantation research. The UpCell™ surface enables harvesting of cell sheets and creation of 3D tissue models held together by normal cell junctions and extracellular matrix deposited by the cells.
- No trypsinisation - cell surface proteins are preserved
- High cell viability without scraping
- Releases adherent cells by reduction of temperature of the cell culture
- Requires minimal hands-on time
Specifications
- Empf. Arbeitsvolumen:3,0 ml
- Bezeichnung:6-Well-Multischale, UpCell™, belüftet
- Seq:01
- Kulturfläche:9,6 cm²
Accessories
Product Information
- Catalog #BezeichnungAvailabilityPrice
- NUNC174922Membrane supports for Nunc™ 12-well multidishes with UpCell™ surfaceLog in for availability1 * 12 ST - SFr. 60.00