Bottle pourers
Bottle pourer, PTFE, blue, for GL 45 screw neck bottles
Katalognr. COWI028.045BLUE
Anbieter: COWIE
Direkt zu
Product Details & Documents
Manufactured in pure, FDA approved, PTFE.
- Totally chemically resistant and inert
- To fit GL45, 38, 32 and 30 screw neck bottles
- State cap colour required, blue, red or green
Permits direct pouring in a safe and consistent manner.
Technische Daten
- Typ:for GL 45 screw neck bottles
- Bezeichnung:Bottle pourer, PTFE, blue