Pipette tips
Bevelled, non-graduated, 1 - 200 µl, -, 50 mm, Gelb, 96 tips/rack; 10 racks/case
Katalognr. ALPCLW6360
Anbieter: Alpha Laboratories
Direkt zu
Product Details & Documents
These tips are manufactured from medical grade PP in modern production facilities with stringent quality controls at every stage.
- Tips are lot tested and certified for absence of detectable human DNA and detectable endotoxins (pyrogens)
- Tips are tested and certified for absence of heavy metals, detectable DNase and RNase
- With graduations to visually check volume
- Suitable for all major pipettor brands
Technische Daten
- Farbcode:Gelb
- Verpackt:96 tips/rack; 10 racks/case
- Steril:-
- Volumen:1 - 200 µl
- Länge:50 mm
- Seq:04
- Spitzentyp:Bevelled, non-graduated