Kühler, Air
500 mm, 19/26 NS
Catalog # WITG1.782.050
Supplier: witeg Labortechnik
Jump to:
Product Details & Documents
Borosilikatglas 3.3
- Sehr gute chemische Beständigkeit
- Hohe Temperaturbeständigkeit
- Minimale Wärmeausdehnung, dadurch bedingt hohe Temperaturwechselbeständigkeit
Mit Normschliffkonus.
Certifications:
Ordering information:
Delivery information:
Packaging:
Caution:
Specifications
- Kern:19/26 NS
- Mantellänge:500 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.