D(+)-Arabitol ≥99%
Glass bottle for solids, 25 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 99% CAS No.: 488-82-4 Molecular Formula: C5H12O5 Formula Weight: 152.15 Melting Point: 100-104° Beilstein Reference: 1720520 Merck Reference: 14,762 Optical Rotation: +132° (c=0.4 in acidified molybdate solution) MDL No.: MFCD00004709
Synonyme: D-Arabitol, (2R,4R)-Pentane-1,2,3,4,5-pentaol
- CAS Nummer:488-82-4
- Formel:C₅H₁₂O₅
- MDL-Nummer:MFCD00004709
- Molekulargewicht:152.15 g/mol
- Merck-Index:14,762
- Schmelzender Platingehalt:100…104 °C
- Lagertemperatur:Raumtemperatur
- EINECS:207-686-2
Documents
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:25 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



