L(-)-Carnitin ≥99%
Glasflasche für Feststoffe, 10 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 99+% CAS No.: 541-15-1 Molecular Formula: C7H15NO3 Formula Weight: 161.20 Melting Point: ca 190° dec. Beilstein Reference: 4292315 Merck Reference: 14,1849 MDL No.: MFCD00038747
Synonyme: Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate
- CAS Nummer:541-15-1
- Formel:C₇H₁₅NO₃
- MDL-Nummer:MFCD00038747
- Molekulargewicht:161.20 g/mol
- Merck-Index:14,1849
- Schmelzender Platingehalt:∼ 190 °C (Zers.)
- Schmelzender Platingehalt:∼ 190 °C (Zersetzung)
- Lagertemperatur:Raumtemperatur
- EINECS:208-768-0
Documents
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:10 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



