Interchangeable blocks for block heaters, digital, SB1
Interchangeable block, suitable only for use with dual block model, 96 well plate, skirted or non-skirted or 4 slides, —
Katalognr. JENC357-138
Anbieter: JENCONS SCIENTIFIC
Interchangeable blocks for block heaters, digital, SB1
Katalognr. JENC357-138
Anbieter: JENCONS SCIENTIFIC
Direkt zu
Product Details & Documents
Interchangeable blocks are made of high grade, non porous, anodised aluminium. Except the blocks used for plates and slides, all other blocks include positions for a block lifter and calibration thermometer.
Technische Daten
- Anz. Bohrungen:—
- Bezeichnung:Interchangeable block, suitable only for use with dual block model
- Seq:17
- Für:96 well plate, skirted or non-skirted or 4 slides