Interchangeable blocks for block heaters, digital, SB1
Interchangeable block, 20 mm Ø tubes, 6
Catalog # JENC357-134
Supplier: JENCONS SCIENTIFIC
Interchangeable blocks for block heaters, digital, SB1
Catalog # JENC357-134
Supplier: JENCONS SCIENTIFIC
Jump to:
Product Details & Documents
Interchangeable blocks are made of high grade, non porous, anodised aluminium. Except the blocks used for plates and slides, all other blocks include positions for a block lifter and calibration thermometer.
Specifications
- Anz. Bohrungen:6
- Bezeichnung:Interchangeable block
- Seq:13
- Für:20 mm Ø tubes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.