Interchangeable blocks for block heaters, digital, SB1
Interchangeable block, 10 mm Ø tube, 20
Katalognr. JENC357-128
Anbieter: JENCONS SCIENTIFIC
Interchangeable blocks for block heaters, digital, SB1
Katalognr. JENC357-128
Anbieter: JENCONS SCIENTIFIC
Direkt zu
Product Details & Documents
Interchangeable blocks are made of high grade, non porous, anodised aluminium. Except the blocks used for plates and slides, all other blocks include positions for a block lifter and calibration thermometer.
Technische Daten
- Anz. Bohrungen:20
- Bezeichnung:Interchangeable block
- Seq:07
- Für:10 mm Ø tube