Pycnometer with thermometer, adjusted
50 ml, 10/19
Katalognr. HECH42572332
Anbieter: GLASWARENFABRIK KARL HECHT
Direkt zu
Product Details & Documents
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
- With ground-in capillary stopper
- Very good chemical resistance
- Very good temperature resistance
It includes pycnometer bottle and ground-in thermometer joint size 10/19 with blue filling.
Zertifizierung: DIN ISO 3507
Dokumente
HECH42572016_TDS_EN HECH42572032_TDS_EN
Technisches Datenblatt
Technische Daten
- NS:10/19
- Volumen (nom.):50 ml
- Cat. No.:HECH42572332