Cork borer sets
Cork borers, set of 6, 5,0 - 11,0 mm
Katalognr. HECH41593006
Anbieter: GLASWARENFABRIK KARL HECHT
Direkt zu
Product Details & Documents
These cork borers are made of nickel-plated brass tubes with handles.
Technische Daten
- Bohrung Ø:5,0 - 11,0 mm
- Bezeichnung:Cork borers, set of 6
- Seq:01