Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Cover glasses | Avantor

Cover glasses

Deckgläser, 0,17 ±0,01 mm, 25 mm
Katalognr. HECH92101401190
Cover glasses
Katalognr. HECH92101401190
Cover glasses
Cover glasses

Deckgläser, 0,17 ±0,01 mm, 25 mm

Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1.000 ST
Product Details & Documents

High spectral transmission, excellent flatness, optimum resistance to chemicals.

  • Free from colour distortion borosilicate glass D 263™ M
  • Excellent quality: Low levels of inclusions, striae, bubbles and streaks

 : ISO 8255

 : In tight-closing plastic boxes for protection against damp and climatic influences.

Technische Daten
  • Ø:
    25 mm
  • Bezeichnung:
    Deckgläser
  • Seq:
    02
  • Stärke:
    0,17 ±0,01 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.