Dissecting scissors
Scharf/scharf, 155 mm
Katalognr. MART11-300-15
Anbieter: KLS Martin Group
Direkt zu
Product Details & Documents
Dissecting scissors made of stainless steel. One model features a TC Gold finish.
- TC Gold finish: Tungsten Carbide inlays provide an exceptionally long service life
- Straight
Technische Daten
- Typ:Scharf/scharf
- Länge:155 mm