Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Notepads, microplate wells
Mikroplatte 48-Well-Pad, 127×178 mm
Katalognr. 613-1456
Anbieter:   DIVERSIFIED BIOTECH
Notepads, microplate wells
Katalognr. 613-1456
Notepads, microplate wells
Notepads, microplate wells
Mikroplatte 48-Well-Pad, 127×178 mm
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
These pads with the layout of a 24-, 48- or 96-well microtiter plate provide a fast and easy way to keep accurate records of samples in a microplate. The adhesive backing allows the researcher to affix individual sheets into a notebook.
  • With adhesive backing for attaching individual sheets
  • Encourages accurate record keeping
Supplied with 50 pads of 127×178 mm.
Technische Daten
  • Bezeichnung:
    Mikroplatte 48-Well-Pad, 127×178 mm
  • Seq:
    02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.