Trypsin Protease, HyClone™
Trypsin, 0,25% 1X, with 2,5 g porcine trypsin in HBSS with 1 g/l EDTA, without calcium, magnesium (liquid), 100 ml
Anbieter: HyClone products (Cytiva)
Trypsin Protease, HyClone™
Katalognr. HYCLSV30031.01
Anbieter: HyClone products (Cytiva)
CAS Nummer:
Direkt zu
Product Details & Documents
Dissociation reagents, such as trypsin, are used to detach and disaggregate adherent cells from the surface or substrate to which they are attached, as well as for dissociating tissues for primary cell culture.
- Derived from porcine pancreas
- Gamma irradiated prior to hydration and filling
- Formulated without calcium and magnesium
Dokumente
Technische Daten
Technische Daten
- VE:100 ml
- Bezeichnung:Trypsin, 0,25% 1X, with 2,5 g porcine trypsin in HBSS with 1 g/l EDTA, without calcium, magnesium (liquid)