Organic reference standards, ethanol FS
Ethanol standard FS, 2,0 mg/ml, Reference standard, 10×1 ml, Ampoules, 10 mL
Catalog # DIAS109209910349
Supplier: DIAGNOSTIC SYSTEMS DIASYS
Organic reference standards, ethanol FS
Catalog # DIAS109209910349
Supplier: DIAGNOSTIC SYSTEMS DIASYS
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Konzentration:2,0 mg/ml
- Packung:Ampoules
- VE:10 mL
- Normen:Reference standard
- Bezeichnung:Ethanol standard FS
- Pk Info:10×1 ml
- Cat. No.:DIAS109209910349
- CAS Nr.:64-17-5
- Bezeichnung:Ethanol
- Formel:C₂H₅OH
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



