Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Transfer pipettes, Samco™ Extra Long | Avantor

Transfer pipettes, Samco™ Extra Long

5,6 ml, 25, 2,3 ml, 5,0 mm, 225 mm, +, 10 per pack
Katalognr. SAMC262-10S
Anbieter:   SAMCO
Transfer pipettes, Samco™ Extra Long
Katalognr. SAMC262-10S
Anbieter:  SAMCO
Transfer pipettes, Samco™ Extra Long
Transfer pipettes, Samco™ Extra Long

5,6 ml, 25, 2,3 ml, 5,0 mm, 225 mm, +, 10 per pack

Preis
SFr. 50.80
Einloggen für Verfügbarkeit
1 * 400 ST
Product Details & Documents

These extra long PE pipettes are ideal for safe and easy sampling from large containers.

  • Excellent for tissue culture applications
  • General purpose pipettes are ideal for routine sampling of serum and plasma
  • Well suited for blood culture and urine analysis

Perfect for tissue culture applications, these pipettes are designed with a built-in pipette bulb that eliminates the need for plugging, packaging, autoclaving and prevents cross-contamination.

Technische Daten
  • Ext.Ø:
    5,0 mm
  • Verpackt:
    10 per pack
  • Steril:
    +
  • Kapazität:
    5,6 ml
  • Länge:
    225 mm
  • Seq:
    02
  • Tropfen pro ml:
    25
  • Saugvolumen:
    2,3 ml
  • Cat. No.:
    SAMC262-10S
  • Umweltzertifizierung:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Umweltfreundlichkeit:
    Umweltschonende Herstellung