Ribbons for IP™ label printing system
IP-R-6000, Farbband-Serie 6000, Harz-Ausführung, Breite 60 mm, 300 m, Schwarz
Katalognr. BRDY066202
Anbieter: Brady
Direkt zu
Product Details & Documents
These IP® printer ribbons are high density ribbon with excellent solvent resistance, good heat resistance and super scratch resistance and have ribbon spindle.
Technische Daten
- Farbe:Schwarz
- Bezeichnung:IP-R-6000, Farbband-Serie 6000, Harz-Ausführung, Breite 60 mm
- Länge:300 m
- Seq:06