Precast gels, Reliant™
Reliant™ DNA Gel, 24-well, 1% SeaKem® Gold agarose with TBE and EtBr, 300≥8000 bp
Katalognr. 733-1576
Anbieter: LONZA
Direkt zu
Product Details & Documents
Reliant™ precast agarose minigels are designed for rapid and reproducible resolution of DNA fragments from 8 bp to 10 kb. Each precast agarose gel is precision cast using our SeaKem® Gold or NuSieve™ 3:1 agarose and are available in a variety of well formats and agarose concentrations.
- Gels are cast from either SeaKem® or NuSieve® agarose and are certified DNase-free
- Gels specifically for RNA separation are guaranteed RNase-free
- Compatible with most minigel chambers
Reliant™ precast agarose minigels are available either pre-stained with ethidium bromide or without stain. The Reliant™ gel System is ideal for analysis of PCR, restriction digest fragments or most cloning needs.
: Supplied in boxes of 20 gels.
Technische Daten
- Bezeichnung:Reliant™ DNA Gel, 24-well, 1% SeaKem® Gold agarose with TBE and EtBr, 300≥8000 bp
- Seq:15