DNA markers
DNA marker, 50 - 1000 bp, 250 μl, 50
Katalognr. LONZ50461
Anbieter: LONZA
Direkt zu
Product Details & Documents
DNA marker is for rapid size estimation of PCR products and restriction fragments. DNA can be visualised by ethidium bromide staining or kinased with radiolabelled ³²P for detection by autoradiography.
Technische Daten
Technische Daten
- VE:50
- Bezeichnung:DNA marker, 50 - 1000 bp, 250 μl