Lames micro-array, HTA™ Slides
Multifunctional microarray slides, 12
Katalognr. 731-3012
Anbieter: Greiner Bio-One
Direkt zu
Product Details & Documents
The HTA™ 1 can be used for the processing of one sample. The HTA™ 12 are partitioned into 12 flat compartments, each with a printable surface of 6×6 mm and a low well rim of 0,5 mm. Twelve samples can be processed simultaneously.
- Multifunctional 3-D amino, 3-D aldehyde or 3-D epoxy surface functionality
- Low well rim of only 0,5 mm, low auto-fluorescence
- Plastic plates for high-throughput microarrays
Combine the advantages of the universal microplate platform with microarrays for diagnostic applications due to identical well geometry compared with a standard 96-well microplate.
Technische Daten
- Anz. Wells:12
- Bezeichnung:Multifunctional microarray slides