Lames micro-array, HTA™ Slides
12, 3-D Amino, 12
Katalognr. 731-3013
Anbieter: Greiner Bio-One
Direkt zu
Product Details & Documents
The HTA™ 1 can be used for the processing of one sample. The HTA™ 12 are partitioned into 12 flat compartments, each with a printable surface of 6×6 mm and a low well rim of 0,5 mm. Twelve samples can be processed simultaneously.
- Multifunctional 3-D amino, 3-D aldehyde or 3-D epoxy surface functionality
- Low well rim of only 0,5 mm, low auto-fluorescence
- Plastic plates for high-throughput microarrays
Combine the advantages of the universal microplate platform with microarrays for diagnostic applications due to identical well geometry compared with a standard 96-well microplate.
Technische Daten
- Anz. Wells:12
- Bezeichnung:12, 3-D Amino
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.