AuftragsverfolgungKontakt
Lames micro-array, HTA™ Slides | Avantor
Lames micro-array, HTA™ Slides
12, 3-D Amino, 12
Katalognr. 731-3013
Anbieter:   Greiner Bio-One
Lames micro-array, HTA™ Slides
Katalognr. 731-3013
Anbieter:  Greiner Bio-One
Lames micro-array, HTA™ Slides
Lames micro-array, HTA™ Slides
12, 3-D Amino, 12
Preis
SFr. 459.00
Einloggen für Verfügbarkeit
1 * 5 ST
Product Details & Documents
The HTA™ 1 can be used for the processing of one sample. The HTA™ 12 are partitioned into 12 flat compartments, each with a printable surface of 6×6 mm and a low well rim of 0,5 mm. Twelve samples can be processed simultaneously.
  • Multifunctional 3-D amino, 3-D aldehyde or 3-D epoxy surface functionality
  • Low well rim of only 0,5 mm, low auto-fluorescence
  • Plastic plates for high-throughput microarrays
Combine the advantages of the universal microplate platform with microarrays for diagnostic applications due to identical well geometry compared with a standard 96-well microplate.
Technische Daten
  • Anz. Wells:
    12
  • Bezeichnung:
    12, 3-D Amino