Poly(α-methylstyrene) analytical standards, for GPC
~427000 Mn, Mw/Mn ~1,02, 250 mg
Katalognr. 81523-250MG
Anbieter: Merck
Poly(α-methylstyrene) analytical standards, for GPC
Katalognr. 81523-250MG
Anbieter: Merck
CAS Nummer:
Direkt zu
Product Details & Documents
Poly(α-methylstyrene) may find applications as a radical producing agent, in the thermal degradation of polystyrene.
Zertifizierung: Exact values can be taken from the accompanying certificate of analysis.
Achtung! Bitte beachten: Storage temperature: 2 to 8 °C. Limited shelf life, expiry date on the label.
Technische Daten
Technische Daten
- VE:250 mg
- Bezeichnung:~427000 Mn, Mw/Mn ~1,02
- Cat. No.:81523-250MG
- M-Clarity Qualitätsstufe:MQ100