Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Benzaldehyd, Supelco®

Benzaldehyd, Supelco®

Glasflasche, 25 mL
Catalog # 09143-25ML-F
Irritant
Supplier:   Merck
Benzaldehyd, Supelco®
Catalog # 09143-25ML-F
Supplier:  Merck
CAS Number:  100-52-7
Benzaldehyd, Supelco®
Benzaldehyd, Supelco®

Glasflasche, 25 mL

CAS Number:  100-52-7
Price
656.00 SFr.
Log in for availability
1 * 25 mL
Product Details & Documents
  • CAS Number:100-52-7
  • Formula:C₆H₅CHO
  • MDL Number:MFCD00003299
  • Molecular Weight:106.12 g/mol
  • Merck Index:13,01057
  • Boiling Pt:179 °C (1013 hPa)
  • Melting Pt:–26 °C
  • Density:1,049 g/cm³ (20 °C)
  • Flash Pt:64 °C
  • Storage Temperature:Raumtemperatur
  • UN:1990
  • ADR:9,III
  •  :M-Clarity™ Qualitätsstufe = MQ100
  • EINECS:202-860-4
Specifications

Specifications

  • Packung:
    Glasflasche
  • VE:
    25 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.