Dichlormethan, Supelco®
Glass bottle, 100 mL
Katalognr. SUPL66742-100ML
Anbieter: Merck
Direkt zu
Product Details & Documents
Synonyme: Methylene chloride, DCM
- CAS Nummer:75-09-2
- Formel:CH₂Cl₂
- MDL-Nummer:MFCD00000881
- Molekulargewicht:84.93 g/mol
- Merck-Index:12,06140
- Siedepunkt:39,8 °C (1013 hPa)
- Schmelzender Platingehalt:–95 °C
- Dichte:1,322 g/cm³ (20 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:200-838-9
- UN:1593
- ADR:6.1,III
Technische Daten
Technische Daten
- Packung:Glass bottle
- VE:100 mL
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



