Nitrobenzol, Supelco®
Glasflasche, 5 mL
Anbieter: Merck
Direkt zu
Product Details & Documents
- CAS Nummer:98-95-3
- Formel:C₆H₅NO₂
- MDL-Nummer:MFCD00007043
- Molekulargewicht:123.11 g/mol
- Merck-Index:13,06621
- Siedepunkt:210,8 °C (1013 hPa)
- Schmelzender Platingehalt:5,7 °C
- Dichte:1,2037 g/cm³ (20 °C)
- Blitz Pt:88 °C
- Lagertemperatur:Raumtemperatur
- UN:1662
- ADR:6.1,II
- :M-Clarity™ Qualitätsstufe = MQ100
- EINECS:202-716-0
Documents
Zertifikate
Suche nach ChargennummerTechnische Daten
Technische Daten
- Packung:Glasflasche
- VE:5 mL
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



