AuftragsverfolgungKontakt
Brenner
Brenner
Radial torches, low flow quartz torch with 1,4 mm ID injector tube
Catalog # VARI2010069690
Supplier:   VARIAN
Brenner
Catalog # VARI2010069690
Supplier:  VARIAN
Brenner
Brenner
Radial torches, low flow quartz torch with 1,4 mm ID injector tube
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Diese Taschenlampen lassen sich leicht anzünden und bleiben an, wodurch Ausfallzeiten reduziert werden. Mit gleichmäßigem und stabilem Plasma für reproduzierbare Ergebnisse.
Certifications:
Accessories information:
Ordering information:
Delivery information:
Packaging:
Specifications
  • Bezeichnung:
    Radial torches, low flow quartz torch with 1,4 mm ID injector tube
  • Seq:
    07
Accessories
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • VARI2010070790
    Spare quartz torch bonnet for radial ICP-OES
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • VARI7210020700
    Torch alignment tool for radial ICP-OES
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • VARI7910051290
    GazFit fittings for one-piece and semi-demountable ICP-OES torches
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • VARI7910051590
    Transfer tube FEP for axial ICP
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • VARI7910051490
    Transfer tube FEP Sturman-Masters spray chamber for radial ICP
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • VARI9910107100
    Kit Torch Fittings
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • VARI3610007300
    Pre-optics cone for Liberty Series axial ICP-OES
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • VARI8110002800
    Esteele cleaning powder, for cleaning interface cone on axially-viewed ICP-OES
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • VARI2010072900
    Spare semi-demountable quartz torch body for radial ICP-OES
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • VARI2010077500
    Quartz injector tube, 0,8 mm
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • VARI2010083400
    Spare semi-demountable quartz torch body for axial ICP-OES
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • VARI9910057000
    Quartz injector tube, 1,4 mm
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • VARI9910057300
    Injector holder kit, includes O-ring
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • VARI9910108200
    O-rings kit