Cary source lamps
Lamp, deuterium, UV, for Cary 4000/5000/6000i
Katalognr. VARI110713990
Anbieter: VARIAN
Direkt zu
Product Details & Documents
Source lamps for Cary spectrophotometers are fully modular and interchangeable. A range of UV, xenon and visible lamps are available.
- Fully modular and compatible with Cary spectrophotometers
The UV, xenon and visible lamps are fully modular, allowing you to select and change between light sources according to your experimental requirements.
Technische Daten
- Bezeichnung:Lamp, deuterium, UV, for Cary 4000/5000/6000i
- Seq:01
Accessories
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- HEWLG9820-60000Loom, deuterium lamp extension, required when installing deuterium lamp mm a Cary ¹/₃/100/300 series UV-VisEinloggen für Verfügbarkeit1 * 1 ST