AuftragsverfolgungKontakt
Micro pipette tips, OMIX® | Avantor
Micro pipette tips, OMIX®
OMIX C18, 10 - 100 µl, 1×96 tips
Katalognr. VARIA57003100
Anbieter:   VARIAN
Micro pipette tips, OMIX®
Katalognr. VARIA57003100
Anbieter:  VARIAN
Micro pipette tips, OMIX®
Micro pipette tips, OMIX®
OMIX C18, 10 - 100 µl, 1×96 tips
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Bond Elut OMIX pipette tips reliably purify and enrich femtomole and picomole levels of peptides and proteins prior to MALDI-TOF or LC/MS/MS. The unique monolithic sorbent technology used in Bond Elut OMIX consistently outperforms other tips by delivering uniform flow and strong analyte-to-surface interactions. Bond Elut OMIX tips bind more peptides to deliver better sequence coverage.
  • Fast, uniform flow maximizes productivity and reproducibility
  • Minimal peptide losses lead to higher recoveries
  • Available in three phases and sizes to deliver better sequence coverage
Technische Daten
  • Verpackt:
    1×96 tips
  • Volumen:
    10 - 100 µl
  • Seq:
    08
  • Spitzentyp:
    OMIX C18