L(-)-Carnitin 99+%
Glasflasche, 1 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate
- CAS Number:541-15-1
- Formula:C₇H₁₅NO₃
- MDL Number:MFCD00038747
- Molecular Weight:161.20 g/mol
- Melting Pt:208…212 °C (Zers.)
- Melting Pt:208…212 °C (Zersetzung)
- Storage Temperature:Kühlschrank
- EINECS:208-768-0
Documents
Specifications
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Infrared spectrum:Conforms
- Solubility:(5 % in water) Clear colorless
- Specific optical rotation:-29° to -32° (20°C, 589 nm) (c=1, H2O)
- Titration with HClO4:≥99.0 %
Specifications
- Packung:Glasflasche
- VE:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



