Triethylcitrat 99%
Kst.-Flasche, 1 kg
: Triethyl 2-hydroxypropane-1,2,3-tricarboxylate, Hydroxytricarballylic acid triethyl ester, Citric acid triethyl ester
- :77-93-0
- :C₁₂H₂₀O₇
- :MFCD00009201
- :276.29 g/mol
- :294 °C (760 mmHg)
- :–46 °C
- :1,136 g/cm³
- :151 °C
- :Raumtemperatur
- :201-070-7
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4400 to 1.4430 (20°C, 589 nm)
- GC:≥98.5 %
- Color scale:≤50 APHA
- Packung:Kst.-Flasche
- VE:1 kg
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



