Methyldimethoxyacetat 98%
Glasflasche, 5 g
: Methyl 2,2-dimethoxyacetate
- :89-91-8
- :C₅H₁₀O₄
- :MFCD00008484
- :134.13 g/mol
- :160…163 °C (760 mmHg)
- :1,09 g/cm³
- :63 °C
- :Kühlschrank
- :201-950-0
Documents
ACRO41418
SDS
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4040 to 1.4070 (20°C, 589 nm)
- GC:≥97.5 %
- Packung:Glasflasche
- VE:5 g



